Skip to Content
Merck
1-Boc-hexahydro-1,4-diazepine 98%

1-Boc-hexahydro-1,4-diazepine

1-Boc-hexahydro-1,4-diazepine 98%
Synonym(s):
1-Boc-homopiperazine, tert-Butyl homopiperazine-1-carboxylate
Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing