1,4-Sorbitan
Synonym(s):
1,4-Anhydro-D-glucitol, 1,4-Anhydro-D-sorbitol, Arlitan, 1,4-Sorbitan
Empirical Formula (Hill Notation):
C6H12O5
CAS Number:
Molecular Weight:
164.16
Products
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing