
1,4-Butanediol-2,2,3,3-d4

Linear Formula:
HOCH2(CD2)2CH2OH
CAS Number:
Molecular Weight:
94.15
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing