
1,1,2,2-Tetrachloroethane-d2

Synonym(s):
1,2-Dideutero-1,1,2,2-tetrachloroethane, 1,1,2,2-Tetrachloroethane-D2
Linear Formula:
Cl2CDCDCl2
CAS Number:
Molecular Weight:
169.86
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing