
1,1,1,3,3,3-Hexakis(dimethylamino)diphosphazenium tetrafluoroborate

Synonym(s):
Bis[tris(dimethylamino)phosphoranylidene]ammonium tetrafluoroborate, Phosphazenium tetrafluoroborate P2-BF4
Linear Formula:
[[(CH3)2N]3P=]2N(BF4)
CAS Number:
Molecular Weight:
427.21
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing