Skip to Content
Merck
All Photos(1)

Key Documents

1581005

USP

Pseudoephedrine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(+)-Pseudoephedrine hydrochloride, (+)-ψ-Ephedrine hydrochloride, (1S,2S)-2-Methylamino-1-phenylpropanol hydrochloride, d-Isoephedrine hydrochloride, d-Pseudoephedrine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH·HCl
CAS Number:
Molecular Weight:
201.69
Beilstein:
3915112
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

pseudoephedrine

manufacturer/tradename

USP

mp

185-188 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CN[C@@H](C)[C@@H](O)c1ccccc1

InChI

1S/C10H15NO.ClH/c1-8(11-2)10(12)9-6-4-3-5-7-9;/h3-8,10-12H,1-2H3;1H/t8-,10+;/m0./s1

InChI key

BALXUFOVQVENIU-KXNXZCPBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pseudoephedrine hydrochloride USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Pseudoephedrine Hydrochloride Tablets
  • Pseudoephedrine Hydrochloride Oral Solution
  • Acetaminophen and Pseudoephedrine Hydrochloride Tablets
  • Ibuprofen and Pseudoephedrine Hydrochloride Tablets
  • Pseudoephedrine Hydrochloride Extended-Release Tablets
  • Pseudoephedrine Hydrochloride Extended-Release Capsules
  • Guaifenesin and Pseudoephedrine Hydrochloride Capsules

Biochem/physiol Actions

Non-selective adrenergic agonist; decongestant

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service