Skip to Content
Merck
All Photos(1)

Documents

1097603

USP

Cefazolin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Cephazolin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N8O4S3
CAS Number:
Molecular Weight:
454.51
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cefazolin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C14H14N8O4S3/c1-6-17-18-14(29-6)28-4-7-3-27-12-9(11(24)22(12)10(7)13(25)26)16-8(23)2-21-5-15-19-20-21/h5,9,12H,2-4H2,1H3,(H,16,23)(H,25,26)/t9-,12-/m1/s1

InChI key

MLYYVTUWGNIJIB-BXKDBHETSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cefazolin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Cefazolin for Injection
  • Cefazolin in Dextrose Injection
  • Cefazolin Sodium

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Osler et al.
Diseases of the colon and rectum, 29(2), 140-143 (1986-02-01)
The seventh case of probable cefazolin-induced pseudomembranous colitis is reported. Perforation of the colon necessitated sigmoid resection. The postoperative course was protracted, and illustrates the difficulty of managing advanced pseudomembranous colitis when the oral route of antibiotic administration is not
Cheng-Kun Wu et al.
PloS one, 8(4), e61666-e61666 (2013-05-01)
Antibiotic prophylaxis with norfloxacin, intravenous ciprofloxacin, or ceftriaxone has been recommended for cirrhotic patients with gastrointestinal hemorrhage but little is known about intravenous cefazolin. This study aimed to compare the outcome of intravenous cefazolin and ceftriaxone as prophylactic antibiotics among
Nagaraju Konda et al.
Optometry and vision science : official publication of the American Academy of Optometry, 91(1), 47-53 (2013-11-12)
To investigate the utility of additional microbial analyses to detect the causative microorganism of microbial keratitis and to determine the strength of association between clinical variables. This retrospective study included 125 eyes of 123 people presenting with presumed infectious keratitis
Angela Macaluso et al.
The journal of pain : official journal of the American Pain Society, 14(6), 604-612 (2013-06-04)
Repeated injections of the antibiotic ceftriaxone cause analgesia in rodents by upregulating the glutamate transporter, GLT-1. No evidence is available in humans. We studied the effect of a single intravenous administration of ceftriaxone in patients undergoing decompressive surgery of the
John D Turnidge
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 52(7), 917-924 (2011-03-24)
Clinical and Laboratory Standards Institute breakpoints for cefazolin against Enterobacteriaceae that were published in January 2010 have been revised by the Subcommittee on Antimicrobial Susceptibility Testing, based on the examination of recent data about in vitro activity, pharmacokinetic-pharmacodynamic characteristics and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service