Skip to Content
Merck
All Photos(1)

Key Documents

PHL89568

β-Asarone

phyproof® Reference Substance

Synonym(s):

cis-1-Propenyl-2,4,5-trimethoxybenzene, cis-Asarone, cis-Isoasarone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CH=CHCH3
CAS Number:
Molecular Weight:
208.25
Beilstein:
1910605
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

liquid

manufacturer/tradename

PhytoLab

refractive index

n20/D 1.558 (lit.)

density

1.073 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]\C(C)=C(/[H])c1cc(OC)c(OC)cc1OC

InChI

1S/C12H16O3/c1-5-6-9-7-11(14-3)12(15-4)8-10(9)13-2/h5-8H,1-4H3/b6-5-

InChI key

RKFAZBXYICVSKP-WAYWQWQTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service