Skip to Content
Merck
All Photos(1)

Key Documents

92265

Supelco

Chlorphenesin

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11ClO3
CAS Number:
Molecular Weight:
202.63
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

grade

analytical standard

Quality Level

Assay

≥98.0 % (GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-30°C

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service