Skip to Content
Merck
All Photos(1)

Key Documents

818388

TSKgel® SuperQ-5PW (20 µm) HPLC Columns

L x I.D. 2.5 cm × 6 mm, ammonium ion, quaternary phase

Synonym(s):

TSKgel® Anion Exchange HPLC Guardgel Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

product name

TSKgel® SuperQ-5PW Guardgel Kit, phase ammonium ion, quaternary, L × I.D. 2.5 cm × 6 mm, 20 μm particle size

material

PEEK

Agency

suitable for USP L23

product line

TSKgel®

feature

counter ion Cl-

manufacturer/tradename

Tosoh 18388

parameter

≤0.5 M salt concentration
≤20% organic solvent
≤45 °C temperature

technique(s)

HPLC: suitable

L × I.D.

2.5 cm × 6 mm

matrix

polymer particle platform

matrix active group

ammonium ion, quaternary phase

particle size

20 μm

pore size

1000 Å

operating pH

2-12

separation technique

anion exchange

General description

Guardgel Kit for 21.5mmID TSKgel SuperQ-5PW columns
TSKgel Guardgel kits contain:
  • 5 mL Guardgel packing material
  • Column holder, consisting of external reducing union, cap, frit, 2 ferrules
  • Column, 25 mm x 9.5 mm OD x 6 mm ID, 2 μm frit at 10 mm depth, 1/16" exit
  • 2 Replacement frits
  • Capillary tubing, 5 cm x 1/16" OD x 0.01" ID
  • Nuts and ferrules, 10-32 threads for 1/16" tubing, 2 each
  • All hardware is 316 stainless steel

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service