Skip to Content
Merck
All Photos(1)

Key Documents

55205

Supelco

Supelguard HPLC Guard Cartridge Holder

Direct-Connect, for use with Supelguard cartridge (2 cm L. x 3 to 4.6 mm I.D.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
Pricing and availability is not currently available.

Product Name

Supelguard Guard Cartridge Holder, Direct-Connect, for use with Supelguard cartridge (2 cm L. x 3 to 4.6 mm I.D.)

material

stainless steel housing

fitting

female for 3.0, 4.0, or 4.6 mm I.D. Supelco brand HPLC columns
10-32 female Valco

compatibility

for use with Supelguard cartridge (2 cm L. x 3 to 4.6 mm I.D.)

Looking for similar products? Visit Product Comparison Guide

General description

Connects guard column directly to a modular analytical column.

Footnote

For 3.0, 4.0 and 4.6mm I.D. Supelco columns.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service