The caps and the vial can be autoclaved, but not the septa.
27180
Vials, screw top with phenolic open-top cap, pre-assembled
volume 40 mL, clear glass vial, O.D. × H 29 mm × 82 mm, tan PTFE/silicone septum
About This Item
Recommended Products
material
black phenolic cap (open-top)
clear glass vial
tan PTFE/silicone septum
feature
closure type black phenolic hole cap
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H
29 mm × 82 mm
volume
40 mL
fitting
thread for 24-400
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
related product
replacement part
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Are the 27180 Vials, screw top with a phenolic open-top cap, pre-assembled volume 40 mL, clear glass vial, O.D. × H 29 mm × 82 mm, tan PTFE/silicone septum, autoclavable?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service