Skip to Content
Merck
All Photos(14)

Key Documents

ZRB1146

Sigma-Aldrich

Anti-Calponin 1 Antibody, clone 9D14, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

9D14, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 33.17 kDa
observed mol wt ~33 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable (peptide)

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CNN1(1264)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 9D14 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Calponin-1. It targets an epitope within 13 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 13 amino acids from the C-terminal region of human Calponin-1.

Application

Anti-Calponin 1, clone 9D14 ZooMAb, Cat. No. ZRB1146, is a recombinant rabbit monoclonal antibody that specifically targets Calponin 1 and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Calponin 1 in human skeletal muscle.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Calponin 1 in human uterus and mouse colon tissue sections.

Note: Actual optimal working dilutions must be determined by the end user as specimens and experimental conditions may vary.

Target description

Calponin-1 (UniProt: P51911; also known as Basic calponin, Calponin H1 smooth muscle) is encoded by the CNN1 gene (Gene ID:1264) in human. Calponin is an actin filament-associated regulatory protein that is expressed in smooth muscle and non-muscle cells of vertebrates. Three isoforms of Calponin have been reported that inhibit actin-activated myosin ATPase activity and stabilize the actin cytoskeleton. Calponin 1 is specifically expressed in smooth muscle cells is involved in fine-tuning of smooth muscle contractility. Calponin 2 is found in multiple cell types, including developing smooth muscles and Calponin 3 is found in the neurons, astrocytes and glial cells. Calponin 1 can bind to actin, calmodulin, troponin C, and tropomyosin. Calponin-1 (and other calponins) has a calponin homology (CH) domain at its N-terminal (aa 28-131) that can function either as a part of actin-binding site or serve as a regulatory structure for actin-binding activity. It also has three calponin-like domains (aa 164-189; 204-229; and 243-268) next to the C-terminal region that are conserved in all calponins. The C-terminal segment varies among different calponins and has differentiated effects on weakening the binding of calponin to F-actin. Deletion of the C-terminal tail segment is shown to enhance the actin-binding and bundling activities of calponins. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.(Ref.: Liu, R., and Jin, J-P (2016). Gene 585(1); 143-153).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose. Normal appearance is a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuki Kamiya et al.
PloS one, 16(4), e0249367-e0249367 (2021-04-23)
Dermal fibroblast aging contributes to aging-associated functional defects in the skin since dermal fibroblasts maintain skin homeostasis by interacting with the epidermis and extracellular matrix. Here, we found that puerarin, an isoflavone present in Pueraria lobata (Kudzu), can prevent the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service