Key Documents
Z743543
Microscope slide wells, 2-wells
well diam. × depth 1 cm × 0.25 mm, pack of 25
About This Item
UNSPSC Code:
41121800
NACRES:
MA.01
Pricing and availability is not currently available.
packaging
pack of 25
manufacturer/tradename
Diversified Biotech MIWE-1000
well diam. × depth
1 cm × 0.25 mm
General description
Pre-cut, adhesive well stickers that firmly adhere to microscope slides. Examine specimens with or without cover slips.
Features and Benefits
  • Convenient - Pre-cut, firmly adhere to microscope slides and enable specimen examination with or without cover slips.
  • Multiple Applications - Examine or count tissue culture cells, yeast, bacteria and other microorganisms.
  • Versatile - Compatible with different styles of microscopes (conventional, inverted or dissecting).
  • Chemically Inert - Made of polyester with an acrylic adhesive. Microscope Slide-Wells are inert and stable under most laboratory conditions.
Choose from one of the most recent versions:
Certificates of Analysis (COA)