Skip to Content
Merck
All Photos(1)

Documents

SML3998

Sigma-Aldrich

NVP-AUY922

new

≥98% (HPLC)

Synonym(s):

5-(2,4-Dihydroxy-5-isopropyl-phenyl)-4-(4-morpholin-4-ylmethyl-phenyl)-isoxazole-3-carboxylic acid ethylamide, 5-[2,4-Dihydroxy-5-(1-methylethyl)phenyl]-N-ethyl-4-[4-(4-morpholinylmethyl)phenyl]-3-isoxazolecarboxamide, AUY922, Luminespib, VER 52296, VER-52296, VER52296

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H31N3O5
CAS Number:
Molecular Weight:
465.54
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=NOC(C2=C(O)C=C(O)C(C(C)C)=C2)=C1C3=CC=C(CN4CCOCC4)C=C3)NCC

Biochem/physiol Actions

High-affinity, potent and selective heat shock protein 90 (HSP90α/β) inhibitor with anti-cancer efficacy in cultures and in vivo.



NVP-AUY922 (Luminespib; VER-52296) is a high-affinity (HSP90β Kd = 1.7 nM), potent and selective heat shock protein 90 inhibitor (HSP90α/β Ki = 9.0/8.2 nM, HSP90α/β IC50 = 7.8/21 nM) that exhibits anti-proliferation potency in cancer cultures (GI50 from 2 to 40 nM among 29 cancer lines) and anti-tumor efficacy in vivo (50-75 mg/kg/day i.p.) by inducing cancer cells G1-G2 arrest and apoptosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brittany Epp-Ducharme et al.
Scientific reports, 11(1), 11103-11103 (2021-05-29)
The heat shock protein 90 inhibitor, luminespib, has demonstrated potent preclinical activity against numerous cancers. However, clinical translation has been impeded by dose-limiting toxicities that have necessitated dosing schedules which have reduced therapeutic efficacy. As such, luminespib is a prime
Suzanne A Eccles et al.
Cancer research, 68(8), 2850-2860 (2008-04-17)
We describe the biological properties of NVP-AUY922, a novel resorcinylic isoxazole amide heat shock protein 90 (HSP90) inhibitor. NVP-AUY922 potently inhibits HSP90 (K(d) = 1.7 nmol/L) and proliferation of human tumor cells with GI(50) values of approximately 2 to 40
Michael Rugaard Jensen et al.
Breast cancer research : BCR, 10(2), R33-R33 (2008-04-24)
Heat shock protein 90 (HSP90) is a key component of a multichaperone complex involved in the post-translational folding of a large number of client proteins, many of which play essential roles in tumorigenesis. HSP90 has emerged in recent years as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service