Skip to Content
Merck
All Photos(1)

Key Documents

SML3179

Sigma-Aldrich

Homer

≥95% (HPLC)

Synonym(s):

6-Hydroxy-N-(4’-((5-(((S)-1-((2S,4R)-4-hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-5-oxopentyl)carbamoyl)-4-(4-methylpiperazin-1-yl)-[1,1’-biphenyl]-3-yl)-4-(trifluoromethyl)-nicotinamide, N-(4′-(5-((S)-1-((2S,4R)-4-Hydroxy-2-(4-(4-methylthiazol-5-yl)benzylcarbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-ylamino)-5-oxopentylcarbamoyl)-4-(4-methylpiperazin-1-yl)biphenyl-3-yl)-6-oxo-4-(trifluoromethyl)-1,6-dihydropyridine-3-carboxamide, WD40 repeat domain protein 5 (WDR5) degrader 8g, WDR5 Degrader 8g, WDR5 PROTAC 8g

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H60F3N9O7S
CAS Number:
Molecular Weight:
1012.15
UNSPSC Code:
12352107
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN1CCN(CC1)C2=C(NC(C3=CNC(C=C3C(F)(F)F)=O)=O)C=C(C4=CC=C(C(NCCCCC(N[C@@H](C(C)(C)C)C(N5C[C@H](O)C[C@H]5C(NCC6=CC=C(C=C6)C7=C(C)N=CS7)=O)=O)=O)=O)C=C4)C=C2

Biochem/physiol Actions

Homer (WDR5 Degrader 8g) is a cell-permeable highly efficacious and potent WDR5-selective heterobifunctional degrader composed of WDR5 antagonist OICR-9429 analog plus a butyl chain spacer linked to an E3 ubiquitin ligase von Hippel-Lindau (VHL)-binding ligand that effectively depletes WDR5 in various cancer cell lines (Dmax = 58% and DC50 = 53 nM in MV4-11WDR5-HiBiT cells, 24 h). Homer is shown to dose-dependently induce WDR5 ubiquitylation and proteasomal degradation (Kd = 18 nM; IC50 cellular & lysate = 13.6 & 2.15 μM, respectively, NanoBRET assay).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anja Dölle et al.
Journal of medicinal chemistry, 64(15), 10682-10710 (2021-05-14)
Histone H3K4 methylation serves as a post-translational hallmark of actively transcribed genes and is introduced by histone methyltransferase (HMT) and its regulatory scaffolding proteins. One of these is the WD-repeat-containing protein 5 (WDR5) that has also been associated with controlling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service