Skip to Content
Merck
All Photos(1)

Documents

SML1637

Sigma-Aldrich

Hexaminolevulinate hydrochloride

≥98% (HPLC)

Synonym(s):

5-Aminolevulinic acid hexyl ester HAL hydrochloride, 5-Aminolevulinic acid hexyl ester hydrochloride, HAL, Hexvix, Hexyl 5-aminolevulinate hydrochloride, P 1206, n-Hexyl 5-aminolevulinate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21NO3 · HCl
CAS Number:
Molecular Weight:
251.75
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

room temp

SMILES string

CCCCCCOC(CCC(CN)=O)=O.[H]Cl

InChI

1S/C11H21NO3.ClH/c1-2-3-4-5-8-15-11(14)7-6-10(13)9-12;/h2-9,12H2,1H3;1H

InChI key

LZYXPFZBAZTOCH-UHFFFAOYSA-N

Biochem/physiol Actions

Hexaminolevulinate (HAL) hydrochloride is a hexyl ester of 5-Aminolevulinic acid (ALA), a precursor to porphyrins including the photosensitizer protoporphyrin IX (PPIX). Hexaminolevulinate is more lipophilic than ALA, and is 50–100 times more efficient than ALA at inducing PpIX, which accumulates preferentially in neoplastic cells. Hexaminolevulinate administration followed by light activation has been used for fluorescence cystoscopy to enhance detection of bladder cancer, in particular carcinoma in situ (CIS). Hexaminolevulinate is also being studied for use in cancer photodynamic therapy. Hexaminolevulinate followed by irradiation produced DNA oxidative damage and apoptosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service