Skip to Content
Merck
All Photos(1)

Key Documents

SML1342

Sigma-Aldrich

API-1

Synonym(s):

4-Amino-5,8-dihydro-5-oxo-8-b-D-ribofuranosyl-pyrido[2,3-d]pyrimidine-6-carboxamide, NSC 177223, NSC177223

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15N5O6
CAS Number:
Molecular Weight:
337.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

O=C1C(C(N)=O)=CN([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C3=C1C(N)=NC=N3

InChI

1S/C13H15N5O6/c14-10-6-7(20)4(11(15)23)1-18(12(6)17-3-16-10)13-9(22)8(21)5(2-19)24-13/h1,3,5,8-9,13,19,21-22H,2H2,(H2,15,23)(H2,14,16,17)/t5-,8-,9-,13-/m1/s1

InChI key

SPBWHPXCWJLQRU-FITJORAGSA-N

Biochem/physiol Actions

API-1 is a potent and specific Akt/PKB inhibitor. API-1 binds to pleckstrin homology (PH) domain of Akt and blocks Akt membrane translocation. The compound inhibits all three members of the family Akt1/PKBa, Akt2/PKBb and Akt3/PKBg. API-1 does not inhibit the activation of PKC, SGK, PKA, STAT3, Erk-1/2 or JNK.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juhua Zhuang et al.
Oncology research, 26(8), 1257-1265 (2018-02-22)
Hepatocellular carcinoma (HCC) as one of the most refractory cancers leads to high mortality worldwide. Long noncoding RNAs have been widely acknowledged as important biomarkers and therapeutic targets in HCC. In this study, we investigated the effects of long noncoding
Joshua R Huot et al.
Cells, 10(7) (2021-08-08)
It has been demonstrated that inhibiting Notch signaling through γ-secretase inhibitor (GSI) treatment increases myogenesis, AKT/mTOR signaling, and muscle protein synthesis (MPS) in C2C12 myotubes. The purpose of this study was to determine if GSI-mediated effects on myogenesis and MPS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service