Skip to Content
Merck
All Photos(1)

Key Documents

SMB01073

Sigma-Aldrich

Eurycomanone

≥90% (LC/MS-ELSD)

Synonym(s):

Pasakbumin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O9
CAS Number:
Molecular Weight:
408.40
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

408.4

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

InChI

1S/C20H24O9/c1-7-4-10(21)13(23)17(3)9(7)5-11-18-6-28-20(27,16(17)18)12(22)8(2)19(18,26)14(24)15(25)29-11/h4,9,11-14,16,22-24,26-27H,2,5-6H2,1,3H3/t9-,11+,12+,13+,14-,16+,17+,18+,19-,20-/m0/s1

InChI key

UCUWZJWAQQRCOR-OKNZMGBLSA-N

General description

Eurycomanone, also known as Pasakbumin A, is a bioactive quassinoid natural compound commonly derived from Eurycoma longifolia (Tongkat Ali). Current research indicates that this plant metabolite is an inhibitor and may possess diverse biological activities, including antitumor, antimalarial, and anticancer properties.

Application

Eurycomanone is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Eurycomanone inhibited Jurkat and K562 leukemia cells viability and proliferation without affecting healthy cells and enhanced testosterone steroidogenesis.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shéhérazade Hajjouli et al.
Molecules (Basel, Switzerland), 19(9), 14649-14666 (2014-09-18)
Eurycomanone and eurycomanol are two quassinoids from the roots of Eurycoma longifolia Jack. The aim of this study was to assess the bioactivity of these compounds in Jurkat and K562 human leukemia cell models compared to peripheral blood mononuclear cells
Bin-Seng Low et al.
Journal of ethnopharmacology, 149(1), 201-207 (2013-07-03)
Eurycoma longifolia Jack (Simaroubaceae family), known locally as 'Tongkat Ali' by the ethnic population, is popularly taken as a traditional remedy to improve the male libido, sexual prowess and fertility. Presently, many tea, coffee and carbonated beverages, pre-mixed with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service