Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300042

Sigma-Aldrich

Anti-phospho-AKT1 (pSer473) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AKT antibody produced in rabbit, Anti-MGC99656 antibody produced in rabbit, Anti-PKB antibody produced in rabbit, Anti-PKB-ALPHA antibody produced in rabbit, Anti-v-akt murine thymoma viral oncogene homolog 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~60 kDa

species reactivity

mouse, human, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(Q-F-SP-Y-S)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer473)

Gene Information

human ... AKT1(207)

Immunogen

Peptide sequence around phosphorylation site of serine 473 (Q-F-S(p)-Y-S), according to the protein AKT1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

General protein kinase capable of phosphorylating several known proteins. Phosphorylates TBC1D4. Signals downstream of phosphatidylinositol 3-kinase (PI3K) to mediate the effects of various growth factors such as platelet-derived growth factor (PDGF), epidermal growth factor (EGF), insulin and insulin-like growth factor I (IGF-I). Plays a role in glucose transport by mediating insulin-induced translocation of the GLUT4 glucose transporter to the cell surface. Mediates the antiapoptotic effects of IGF-I. Mediates insulin-stimulated protein synthesis by phosphorylating TSC2 at 'Ser-939' and 'Thr-1462', thereby activating mTORC1 signaling and leading to both phosphorylation of 4E-BP1 and in activation of RPS6KB1. Promotes glycogen synthesis by mediating the insulin-induced activation of glycogen synthase.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riley Bergman et al.
Breast cancer research and treatment, 197(2), 307-317 (2022-11-18)
Increased body mass index (BMI) and metabolic syndrome (MS) are associated with increased breast cancer recurrence risk. Whether this is due to intrinsic tumor biology or modifiable factors of the obese state remains incompletely understood. Oncotype DX Recurrence Scores of
Yingpinyapat Kittirat et al.
Oncology letters, 15(1), 347-353 (2018-02-02)
The protein 14-3-3ζ contributes important regulatory functions in several cellular processes via binding to phosphorylated serine/threonine residues, which promotes cell cycle progression, cell proliferation and anti-apoptosis in multiple types of cancer. The aim of the present study was to investigate
Yoki Nakamura et al.
Journal of neurochemistry, 131(6), 755-766 (2014-09-02)
Intraplantar injection of 0.4% formalin into the rat hind paw leads to a biphasic nociceptive response; an 'acute' phase (0-15 min) and 'tonic' phase (16-120 min), which is accompanied by significant phosphorylation of extracellular signal-regulated kinase (ERK)1/2 in the contralateral
Supharada Tessiri et al.
PeerJ, 10, e12750-e12750 (2022-01-25)
Genetic alterations in ARID1A were detected at a high frequency in cholangiocarcinoma (CCA). Growing evidence indicates that the loss of ARID1A expression leads to activation of the PI3K/AKT pathway and increasing sensitivity of ARID1A-deficient cells for treatment with the PI3K/AKT
Bárbara Kunzler Souza et al.
Molecular neurobiology, 55(9), 7242-7258 (2018-02-06)
There is an urgent need for advances in the treatment of Ewing sarcoma (EWS), an aggressive childhood tumor with possible neuroectodermal origin. Inhibition of histone deacetylases (HDAC) can revert aberrant epigenetic states and reduce growth in different experimental cancer types.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service