Skip to Content
Merck
All Photos(2)

Key Documents

SAB1400200

Sigma-Aldrich

Anti-PER1 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-MGC88021, Anti-PER, Anti-RIGUI, Anti-hPER

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PER1(5187)

Related Categories

General description

This gene is a member of the Period family of genes and is expressed in a circadian pattern in the suprachiasmatic nucleus, the primary circadian pacemaker in the mammalian brain. Genes in this family encode components of the circadian rhythms of locomotor activity, metabolism, and behavior. Circadian expression in the suprachiasmatic nucleus continues in constant darkness, and a shift in the light/dark cycle evokes a proportional shift of gene expression in the suprachiasmatic nucleus. The specific function of this gene is not yet known. Alternative splicing has been observed in this gene; however, these variants have not been fully described. (provided by RefSeq)

Immunogen

PER1 (NP_002607.1, 1 a.a. ~ 1290 a.a) full-length human protein.

Sequence
MSGPLEGADGGGDPRPGESFCPGGVPSPGPPQHRPCPGPSLADDTDANSNGSSGNESNGHESRGASQRSSHSSSSGNGKDSALLETTESSKSTNSQSPSPPSSSIAYSLLSASSEQDNPSTSGCSSEQSARARTQKELMTALRELKLRLPPERRGKGRSGTLATLQYALACVKQVQANQEYYQQWSLEEGEPCSMDMSTYTLEELEHITSEYTLQNQDTFSVAVSFLTGRIVYISEQAAVLLRCKRDVFRGTRFSELLAPQDVGVFYGSTAPSRLPTWGTGASAGSGLRDFTQEKSVFCRIRGGPDRDPGPRYQPFRLTPYVTKIRVSDGAPAQPCCLLIAERIHSGYEAPRIPPDKRIFTTRHTPSCLFQDVDERAAPLLGYLPQDLLGAPVLLFLHPEDRPLMLAIHKKILQLAGQPFDHSPIRFCARNGEYVTMDTSWAGFVHPWSRKVAFVLGRHKVRTAPLNEDVFTPPAPSPAPSLDTDIQELSEQIHRLLLQPVHSPSPTGLCGVGAVTSPGPLHSPGSSSDSNGGDAEGPGPPAPVTFQQICKDVHLVKHQGQQLFIESRARPQSRPRLPATGTFKAKALPCQSPDPELEAGSAPVQAPLALVPEEAERKEASSCSYQQINCLDSILRYLESCNLPSTTKRKCASSSSYTTSSASDDDRQRTGPVSVGTKKDPPSAALSGEGATPRKEPVVGGTLSPLALANKAESVVSVTSQCSFSSTIVHVGDKKPPESDIIMMEDLPGLAPGPAPSPAPSPTVAPDPAPDAYRPVGLTKAVLSLHTQKEEQAFLSRFRDLGRLRGLDSSSTAPSALGERGCHHGPAPPSRRHHCRSKAKRSRHHQNPRAEAPCYVSHPSPVPPSTPWPTPPATTPFPAVVQPYPLPVFSPRGGPQPLPPAPTSVPPAAFPAPLVTPMVALVLPNYLFPTPSSYPYGALQTPAEGPPTPASHSPSPSLPALPPSPPHRPDSPLFNSRCSSPLQLNLLQLEELPRAEGAAVAGGPGSSAGPPPPSAEAAEPEARLAEVTESSNQDALSGSSDLLELLLQEDSRSGTGSAASGSLGSGLGSGSGSGSHEGGSTSASITRSSQSSHTSKYFGSIDSSEAEAGAARGGAEPGDQVIKYVLQDPIWLLMANADQRVMMTYQVPSRDMTSVLKQDRERLRAMQKQQPRFSEDQRRELGAVHSWVRKGQLPRALDVMACVDCGSSTQDPGHPDDPLFSELDGLGLEPMEEGGGEQGSSGGGSGEGEGCEEAQGGAKASSSQDLAMEEEEEGRSSSSPALPTAGNCTS

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service