Skip to Content
Merck
All Photos(1)

Key Documents

R2383

Sigma-Aldrich

Replica plater for 96 well plate

8 × 6 Array

Synonym(s):

96-well plate replicator, Microplate replicator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

anodized aluminum handle
stainless steel

sterility

non-sterile

size

96 wells

color

none

Looking for similar products? Visit Product Comparison Guide

General description

The prongs are 3/4 inch long and 0.125 inches in diameter. Each device has a semi-circular prong in the upper left corner for unambiguous orientation of the replica plate. All other prongs are circular.

Application

For the efficient transfer of bacterial colonies from 96 well multiwell plates to agar plates or nitrocellulose membranes.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gili Regev-Yochay et al.
Journal of clinical microbiology, 46(2), 747-749 (2007-11-28)
Streptococcus pneumoniae is bactericidal to Staphylococcus aureus in vitro. To determine whether this in vitro effect accounts for the inverse relation between S. pneumoniae and S. aureus colonization reported in previous epidemiologic studies, we compared S. pneumoniae and S. aureus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service