Skip to Content
Merck
All Photos(1)

Documents

MAK507

Sigma-Aldrich

Magnesium Assay Kit

sufficient for 250 colorimetric tests

Synonym(s):

Mg Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

application(s)

cosmetics
food and beverages
food and beverages
pharmaceutical

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes; immunological diseases; cardiovascular diseases; musculoskeletal disorders; neurological disorders

shipped in

ambient

storage temp.

2-8°C

General description

Magnesium (Mg) is one of the most abundant and essential minerals in mammals. Magnesium is involved in more than 300 biochemical reactions in the body and plays important roles in muscle and nerve functions, heart rhythm, immune system and bone formation. Magnesium deficiency may lead to nausea, fatigue, muscle contractions, hypocalcemia and hypokalemia.

Features and Benefits

  • Linear detection range 0.1 mg/dL (41 µM) to 3 mg/dL (1.2 mM) Mg2+
  • Compatible with high-throughput handling systems.
  • The optimized formulation has greatly enhanced reagent and signal stability.
  • Low interference in biological samples, assays can be directly performed in serum and urine samples.

Suitability

The kit is suitable for magnesium determination in serum, urine, food, beverages, water, and soil, as well as for studying the effects of drugs on magnesium metabolism.

Principle

The Magnesium Assay Kit is designed to measure magnesium directly in biological samples without any pretreatment. A calmagite dye in the kit forms a colored complex specifically with magnesium. The intensity of the color, measured at 500 nm, is directly proportional to the magnesium concentration in the sample. The optimized formulation minimizes interference by potential substances.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service