Skip to Content
Merck
All Photos(1)

Documents

I1896

Sigma-Aldrich

Interleukin-13 from mouse

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

IL-13

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

1-10 ng/mL EC50

mol wt

11.5 kDa (predicted; 9 kDa under reducing conditions)

packaging

pkg of 5 μg

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Il13(16163)

Biochem/physiol Actions

Interleukin-13 is a pleiotropic cytokine produced by activated Th2 cells in the mouse and human. IL-13 is secreted mainly as an unglycosylated protein of 132 amino acids with a molecular weight of 10,000 daltons. A 4.3kb DNA fragment of the mouse IL-13 gene was sequenced and occurs as a single copy mapping a chromosome 11. In the human, a 4.6 kb DNA segment of the IL-13 gene occurs as a single copy and maps to chromosome 5. IL-13 induces B cell proliferation and also induces IgE switching. The proliferative activity of human and mouse IL-13 products are tested in culture using the human erythroleukemic cell line TF-1.

Physical form

lyophilized from a 0.2 μm filtered solution in phosphate buffered saline (PBS), pH 7.4, with 50 μg bovine serum albumin (BSA) per 1 μg of cytokine

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luke Hatchwell et al.
Thorax, 70(9), 854-861 (2015-06-26)
Asthma exacerbations represent a significant disease burden and are commonly caused by rhinovirus (RV), which is sensed by Toll-like receptors (TLR) such as TLR7. Some asthmatics have impaired interferon (IFN) responses to RV, but the underlying mechanisms of this clinically
Kihyuk Shin et al.
Clinical and experimental medicine, 15(3), 233-244 (2014-06-14)
The leukotrienes (LTs) enhance allergen- and interleukin (IL)-13-dependent allergic lung inflammatory disease. However, the precise requirement of LTs and the mechanism by which they elicit allergic lung responses remain uncertain. To clarify the involvement of LTs in respiratory allergen- and
Gui Yang et al.
The Journal of biological chemistry, 290(20), 12858-12867 (2015-04-04)
Restoration of the antigen (Ag)-specific immune tolerance in an allergic environment is refractory. B cells are involved in immune regulation. Whether B cells facilitate the generation of Ag-specific immune tolerance in an allergic environment requires further investigation. This paper aims
Arif Yurdagul et al.
Cell metabolism, 31(3), 518-533 (2020-02-01)
Continual efferocytic clearance of apoptotic cells (ACs) by macrophages prevents necrosis and promotes injury resolution. How continual efferocytosis is promoted is not clear. Here, we show that the process is optimized by linking the metabolism of engulfed cargo from initial
C Abrial et al.
British journal of pharmacology, 172(17), 4319-4330 (2015-06-05)
15-Lipoxygenase (15-LOX) activity is associated with inflammation and immune regulation. The objectives of the present study were to investigate the expression of 15-LOX-1 and 15-LOX-2 and evaluate the enzymes' roles in the polarization of human lung macrophages (LMs) in response

Articles

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service