Skip to Content
Merck
All Photos(1)

Key Documents

CA46

95010509, human blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CA46, 95010509

biological source

human blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Expresses surface Immunoglobulin M (IgM) (kappa)and secretes Immunoglobulin M (IgM) which does not bind to protein A.

receptors

Surface Immunoglobulin M (IgM) (kappa)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Burkitt′s Lymphoma

Cell Line Description

Derived from ascites fluid of a patient with Burkitt′s Lymphoma. The cells are Epstein Barr Virus Nuclear Antigen (EBNA) negative.CA46 also expresses surface IgM kappa and secretes IgM which does not bind to protein A. Twelve percent of the cells were shown to express complement receptors whereas Fc receptors were undetectable.

Culture Medium

RPMI 1640 + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures at 5-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service