Skip to Content
Merck
All Photos(1)

Key Documents

A-72

89050908, dog thigh, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

A-72, 89050908

biological source

dog thigh

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Canine Golden Retriever tumour

Cell Line Description

Derived from tumour of thigh (unknown histology) of 8 year old Golden Retriever which had a previous history of mammary adenocarcinoma. Susceptible to canine corona-, canine Ad type I and II, canine herpes, canine parainfluenza and canine parvo- viruses.

Application

Virus studies: Canine corona-, Ad I and II, parainfluenza, herpes and parvo- viruses

Culture Medium

L15 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin; 0.02% EDTA; 37°C. CO2 is detrimental; free gas exchange with atmospheric air necessary.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service