Skip to Content
Merck
All Photos(1)

Key Documents

BT

89031603, bovine nose, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

BT, 89031603

biological source

bovine nose

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Bovine turbinate, BVDV negative

Cell Line Description

Derived from new-born bovine turbinate tissue. Used in propagation of BVDV and IBRV, some strains of parainfluenza 3 virus (P1-3V), some types of bovine adenovirus and enterovirus. Negative for BVDV.

Culture Medium

EMEM (HBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Horse Serum (HS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 3x10,000 cells/cm2 using 0.25% trypsin; 5% CO2;37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service