Skip to Content
Merck
All Photos(1)

Documents

93516

Sigma-Aldrich

Atto Rho6G NHS ester

suitable for fluorescence, ≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

ATTO-TEC GmbH

λ

in ethanol (with 0.1% trifluoroacetic acid)

UV absorption

λ: 533-539 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto Rho6G NHS ester is a new rhodamine dye, based on the well-known laser dye Rhodamine 6G. The new label is functionalized for coupling to biomolecules such as DNA, RNA or proteins. Atto Rho6G NHS ester shows strong absorption, extraordinary high fluorescence quantum yield, high thermal and photo-stability, and very little triplet formation.
After coupling to a substrate Atto Rho6G NHS ester carries a net electric charge of +1.
The label is moderately hydrophilic.

find more information here

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service