Skip to Content
Merck
All Photos(1)

Documents

309M-1

Sigma-Aldrich

Oct-4 (MRQ-10) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

diluted ascites fluid

antibody product type

primary antibodies

clone

MRQ-10, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (309M-14)
vial of 0.5 mL concentrate (309M-15)
bottle of 1.0 mL predilute (309M-17)
vial of 1.0 mL concentrate (309M-16)
bottle of 7.0 mL predilute (309M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

isotype

IgG

control

seminoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... POU5F1(5460)

General description

Oct-4 is a transcription factor that functions in the regulation and maintenance of pluripotency in embryonic stem and primordial germ cells. Oct-4 immunoreactivity has been demonstrated in gonadal and extra-gonadal seminomas, dysgerminomas, and embryonal carcinomas. In addition, the immunohistochemical detection of Oct-4 assists in the evaluation of intratubular germ cell neoplasia (IGCN).
Oct-4 is a transcription factor that maintains and regulates pluripotency in embryonic stem and germ cells. Anti-Oct-4 has shown a very high sensitivity and specificity in seminoma/dysgerminoma, embryonal carcinoma, and the germ cell component of gonadoblastoma by nuclear immunostaining. Clear cell carcinoma may enter the differential diagnosis of dysgerminoma as both may grow in nests or tubules, contain clear cells, and have a prominent inflammatory infiltrate (lymphocytes in dysgerminoma and plasma cells in clear cell carcinoma). In one study that looked at anti-Oct-4 staining in clear cell carcinomas, 4 of 14 tumors were found to be focally positive. In another study, 49 endometrioid carcinomas were Oct-4 negative. Rarely dysgerminoma may have a morphologic appearance that overlaps with sex cord-stromal tumors, especially poorly differentiated Sertoli cell tumors. In two studies however, all sex cord-stromal tumors of the testis and granulosa cell tumors of the ovary were Oct-4 negative.

Quality


IVD

IVD

IVD

RUO

Linkage

Oct-4 Positive Control Slides, Product No. 309S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia M Baker et al.
International journal of gynecological pathology : official journal of the International Society of Gynecological Pathologists, 24(1), 39-55 (2005-01-01)
Immunohistochemistry has become an important tool in the diagnosis of ovarian tumors. This article reviews the role of immunohistochemistry in the differential diagnosis of the three main categories of ovarian tumors, with emphasis on recently developed antibodies. In the surface
Martine Cools et al.
The Journal of clinical endocrinology and metabolism, 91(6), 2404-2413 (2006-04-13)
The purpose of the study was to define the histological origin of gonadoblastomas, allowing the identification of high-risk patients. Sixty paraffin-embedded gonadectomy or gonadal biopsy samples of 43 patients with gonadal dysgenesis were selected from our archives. We studied the
K Biermann et al.
Histopathology, 49(3), 290-297 (2006-08-22)
To compare the suitability of new seminoma markers including transcription factors AP-2gamma, OCT3/4 and M2A for detection of metastatic and extragonadal seminomas with the two well-known markers c-KIT and PLAP. The immunohistochemical distribution of PLAP, c-KIT, M2A, AP-2gamma and OCT3/4
Chien-Jui Cheng et al.
Journal of biomedical science, 14(6), 797-807 (2007-08-09)
Testicular germ cell tumors (TGCTs), comprised of seminomas and non-seminomas, are derived from premalignant and noninvasive intracellular germ cell neoplasias. Among TGCTs, seminomas are believed to resemble a transformed state of primordial germ cells (PGCs) and are known to exhibit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service