Skip to Content
Merck
All Photos(3)

Key Documents

SMB00962

Sigma-Aldrich

Phenylacetyl-L-glutamine

≥95%

Synonym(s):

(S)-5-Amino-5-oxo-2-(2-phenylacetamido)pentanoic acid, N-Phenylacetylglutamine, Phenylacetylglutamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N2O4
CAS Number:
Molecular Weight:
264.28
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26

biological source

synthetic

Quality Level

grade

research grade

Assay

≥95%

form

powder

storage condition

(Store in tight containers in a cool and dry place)

technique(s)

HPLC: suitable

color

off-white

cation traces

C: 56.1—62%

storage temp.

room temp

InChI

1S/C13H16N2O4/c14-11(16)7-6-10(13(18)19)15-12(17)8-9-4-2-1-3-5-9/h1-5,10H,6-8H2,(H2,14,16)(H,15,17)(H,18,19)/t10-/m0/s1

InChI key

JFLIEFSWGNOPJJ-JTQLQIEISA-N

General description

Phenylacetyl-L-glutamine is a primary human metabolite formed from phenylacetate in presence of glutamine in the liver. It is also considered as a gut-microbiome derived uremic toxin. Phenylacetyl-L-glutamine urine levels serve as an effective biomarker for the excretion of nitrogenous waste. Also, peripheral artery disease patients exhibit higher circulating concentration of phenylacetyl-L-glutamine, and high plasma phenylacetyl-L-glutamine levels are associated with cardiovascular disease.

Application

Phenylacetyl-ʟ-glutamine is a versatile compound that finds application in microbiome and metabolomics research.

Features and Benefits

  • Ideal for biochemical, microbiome and metabolomics studies
  • High quality product for research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandi M Azab et al.
Journal of clinical medicine, 9(6) (2020-06-21)
Peripheral artery disease (PAD) is characterized by the atherosclerotic narrowing of lower limb vessels, leading to ischemic muscle pain in older persons. Some patients experience progression to advanced chronic limb-threatening ischemia (CLTI) with poor long-term survivorship. Herein, we performed serum
Lu Dai et al.
Toxins, 12(6) (2020-05-31)
Gut microbial metabolism is not only an important source of uremic toxins but may also help to maintain the vitamin K stores of the host. We hypothesized that sevelamer therapy, a commonly used phosphate binder in patients with end-stage kidney
Ina Nemet et al.
Cell, 180(5), 862-877 (2020-03-07)
Using untargeted metabolomics (n = 1,162 subjects), the plasma metabolite (m/z = 265.1188) phenylacetylglutamine (PAGln) was discovered and then shown in an independent cohort (n = 4,000 subjects) to be associated with cardiovascular disease (CVD) and incident major adverse cardiovascular events (myocardial infarction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service