Skip to Content
Merck
All Photos(1)

Key Documents

NIST606

Trace Elements in Basalt Glass

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

packaging

pkg of 1 g

manufacturer/tradename

NIST®

technique(s)

XRF: suitable

application(s)

glass & ceramic

storage temp.

15-25°C

General description

This Standard Reference Material (SRM) is intended to facilitate development of chemical methods of analysis for trace elements in a natural ferro-magnesian silicate glass intended for use with measurement techniques such as laser ablation inductively coupled plasma mass spectrometry (LA-ICP-MS) and microbeam X-ray fluorescence (μXRF). This SRM can also be used for quality assurance when assigning values to in-house control materials.A unit of SRM 606 consists of a single piece of basalt glass mounted in epoxy. The epoxy mounted material has a cylindrical shape approximately 14 mm tall and 12.5 mm in diameter.

SRM 606_cert

SRM 606_SDS

Other Notes

Certified for the analytes listed below.
Barium (Ba), Cobalt (Co), Chromium (Cr), Copper (Cu), Lanthanum (La), Lead (Pb), Scandium (Sc), Strontium (Sr), Vanadium (V), Yttrium (Y), Zinc (Zn)

Reference/Informational value is provided for the following analytes. Cerium (Ce)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service