Skip to Content
Merck
All Photos(1)

Key Documents

NIST141E

Acetanilide

NIST® SRM®141E

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

form

powder

packaging

pkg of 2 g

manufacturer/tradename

NIST®

technique(s)

GC/MS: suitable
NMR: suitable

application(s)

general analytical

General description

Each unit of SRM 141E contains 2 g of crystalline material.

SRM 141E_cert

SRM 141E_SDS

Application

Acetanilide standard reference material (SRM) is intended for use in validating microchemical procedures, used for the determination of carbon, hydrogen, and nitrogen in organic matter.

Features and Benefits

  • SRM 141E is available with a certificate documenting NIST-certified mass fraction values of acetanilide.
  • Certified values are determined using nuclear magnetic resonance (NMR) spectroscopy and gas chromatography-mass spectrometry (GC-MS).  
  • The NIST certificate is provided with storage and use instructions.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Carbon (C), Hydrogen (H), Nitrogen (N) , Oxygen (O)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

321.8 °F - closed cup

Flash Point(C)

161 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service