Skip to Content
Merck
All Photos(1)

Key Documents

84033

Supelco

Ruthenium standard for AAS

analytical standard, ready-to-use, in hydrochloric acid

Synonym(s):

Ruthenium(III) chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

in hydrochloric acid
1.000 g/L Ru

technique(s)

AAS: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

SMILES string

[Ru]

InChI

1S/Ru

InChI key

KJTLSVCANCCWHF-UHFFFAOYSA-N

Preparation Note

prepared with RuCl3 and HCl (~1 M)

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The determination of ruthenium by atomic absorption spectrophotometry
Montford.B and Scribbs.C.B
Analytica Chimica Acta, 53, 101-108 (1971)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service