Skip to Content
Merck
All Photos(1)

Key Documents

80656

Supelco

Sodium polytungstate solution

~85% in H2O, for separation of minerals

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na6O39W12
CAS Number:
Molecular Weight:
2968.00
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

for separation of minerals

form

solution

composition

Hexasodium dihydrogen-dodecawolframate monohydrate, ≥70-90%

concentration

~85% in H2O

technique(s)

centrifugation: suitable

color

cream white,

pH

3.0 at 4,000 g/l(-20 °C68 °F)

solubility

water: soluble

density

5.470 g/cm3-20 °C (2(68 °F))

suitability

suitable for IR spectroscopy

application(s)

general analytical

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O

InChI

1S/6Na.39O.12W/q6*+1;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;6*-1;;;;;;;;;;;;

InChI key

AZCSOJKJFMWYCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Transparent sodium polytungstate polyoxometalate aquatic shields toward effective X-ray radiation protection: Alternative to lead glasses.: This study explores the development of transparent sodium polytungstate polyoxometalate-based shields for effective X-ray radiation protection. The research demonstrates that these aquatic shields can serve as a non-toxic alternative to traditional lead glasses, offering high transparency and efficient radiation attenuation. This makes them suitable for various applications in medical and industrial X-ray protection (Hashemi et al., 2022).

  • Hybrid of sodium polytungstate polyoxometalate supported by the green substrate for photocatalytic degradation of auramine-O dye.: This paper investigates the photocatalytic properties of sodium polytungstate polyoxometalate supported on a green substrate for the degradation of auramine-O dye. The study highlights the efficiency of this hybrid material in environmental applications, particularly in wastewater treatment, by utilizing photocatalytic processes to break down harmful dyes (Mousavi et al., 2022).

  • Bioinorganic Synthesis of Sodium Polytungstate/Polyoxometalate in Microbial Kombucha Media for Precise Detection of Doxorubicin.: This research presents a novel biosensor developed using sodium polytungstate/polyoxometalate synthesized in a microbial kombucha medium. The biosensor shows high sensitivity and precision in detecting doxorubicin, a chemotherapeutic drug, in biological fluids. This innovative approach enhances the biosensor′s performance, making it suitable for clinical applications in drug monitoring and pharmacokinetics studies (Mousavi et al., 2022).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabine Ring et al.
Journal of immunology (Baltimore, Md. : 1950), 194(8), 3735-3744 (2015-03-18)
Dendritic cells (DC) are one target for immune suppression by regulatory T cells (Treg), because their interaction results in reduced T cell stimulatory capacity and secretion of inhibitory cytokines in DC. We show that DC in the presence of Treg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service