Skip to Content
Merck
All Photos(1)

Key Documents

74632

Sigma-Aldrich

Rebaudioside M

≥95.0% (HPLC)

Synonym(s):

Kaur-16-en-18-oic acid, 13-[(O-β-D-glucopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl)oxy]-, O-β-D-glucopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl ester, (4α)-, Rebaudioside X

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H90O33
CAS Number:
Molecular Weight:
1291.29
UNSPSC Code:
12352201
NACRES:
NA.28

Quality Level

Assay

≥95.0% (HPLC)

form

solid

color

white to off-white

application(s)

food and beverages

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Rebaudioside M is a diterpene glycoside isolated from Stevia rebaudiana leaves. Rebaudioside M is a zero-calorie sweetener and has a clean sweet taste with slight bitter or licorine aftertaste.

Application

<ul>
<li><strong>Acute and two-week effects of neotame, stevia rebaudioside M and sucrose-sweetened biscuits on postprandial appetite and endocrine response in adults with overweight/obesity</strong>: This randomized crossover trial examines how Rebaudioside M impacts appetite regulation and hormonal responses after consumption, providing insights into its application in dietary management and obesity control (Gibbons et al., 2024).</li>
<li><strong>Dynamic characteristics of sweetness and bitterness and their correlation with chemical structures for six steviol glycosides</strong>: Research detailing the sensory profiles of Rebaudioside M among other glycosides, useful for optimizing flavor profiles in sweetener development and food science applications (Tian et al., 2022).</li>
</ul>

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Indra Prakash et al.
Foods (Basel, Switzerland), 3(1), 162-175 (2014-02-27)
This work aims to review and showcase the unique properties of rebaudioside M as a natural non-caloric potential sweetener in food and beverage products. To determine the potential of rebaudioside M, isolated from Stevia rebaudiana Bertoni, as a high potency
Zhi Qiao et al.
The Journal of organic chemistry, 83(19), 11480-11492 (2018-09-06)
With cheap and easily available mixtures of steviol glycosides as starting materials, a practical method for steviol acquisition has been developed, on the basis of which a facile, diversity-oriented, and economic protocol for the synthesis of structurally defined steviol glycosides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service