Skip to Content
Merck
All Photos(1)

Key Documents

72834

Supelco

Acrylamide-d3 Standard solution

~500 mg/L in acetonitrile, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164500
NACRES:
NA.21
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

~500 mg/L in acetonitrile

format

single component solution

storage temp.

2-8°C

SMILES string

NC(=O)C(=C([2H])[2H])[2H]

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)/i1D2,2D

InChI key

HRPVXLWXLXDGHG-FUDHJZNOSA-N

General description

Acrylamide-d3 is isotopically labelled or deuterated acrylamide and is mostly used in quantitative proteomics. It has 3 deuteriums instead of protons making it heavier than acrylamide. It can be combined with two-dimensional polyacrylamide gel electrophoresis.

Application

Acrylamide-d3 may be used as an internal standard during the determination of acrylamide in cocoa and coffee products using liquid chromatography–tandem mass spectrometry.[1]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A modified sample preparation for acrylamide determination in cocoa and coffee products.
Arisseto, Adriana Pavesi, et al.
Food Analytical Methods, 1.1, 49-55 (2008)
Da-Eun Kang et al.
Food science and biotechnology, 29(1), 141-148 (2020-01-25)
The aim of this study was to investigate the formation of furan and acrylamide and to compare antioxidant capacities and volatile compounds in cold or hot brewed coffees. Cold brews were prepared at 5 °C and 20 °C for 12 h. using steeping

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service