Skip to Content
Merck
All Photos(2)

Key Documents

49962

Supelco

Ginkgolic acid C13:0

analytical standard

Synonym(s):

2-Hydroxy-6-tridecylbenzoic acid, 6-Tridecylsalicylic acid, Ginkgoneolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O3
CAS Number:
Molecular Weight:
320.47
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCc1cccc(O)c1C(O)=O

InChI

1S/C20H32O3/c1-2-3-4-5-6-7-8-9-10-11-12-14-17-15-13-16-18(21)19(17)20(22)23/h13,15-16,21H,2-12,14H2,1H3,(H,22,23)

InChI key

VEPUCZUJLKAVNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ginkgolic acid C13:0 is a compound obtained from Ginkgo biloba and is effectively used as an antimicrobial agent against S.mutans.
Ginkgolic acid C13:0 is a compound obtained from Ginkgo biloba and is effectively used as an antimicrobial agent against S.mutans.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: ginkgo

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Dental Caries (2015)
P J Buckle et al.
Agents and actions, 10(4), 361-367 (1980-09-01)
Three novel marine substances, 6-n-tridecylsalicylic acid, flexibilide and dendalone 3-hydroxybutyrate, isolated from a brown alga, a soft coral and a dictyoceratid sponge respectively, were active after oral administration in both acute and chronic animal models of inflammation. 6-n-Tridecylsalicylic acid has
Mengyan Wang et al.
Animals : an open access journal from MDPI, 10(6) (2020-05-30)
In this study, we precisely constructed and transfected the overexpression and interference vectors in BFFs to evaluate the role of DLK1 gene on lipid metabolism in vitro. The expression of of DLK1 in the mRNA and protein level tended to
Dental Caries (2015)
Ping Jiang et al.
Journal of dairy science, 103(7), 6661-6671 (2020-05-04)
The CD44 gene encodes a cell-surface glycoprotein that participates in a variety of biological processes such as cell interactions, adhesion, hematopoiesis, and tumor metastasis. We compared the transcriptome in bovine mammary epithelial cells (bMEC) of Chinese Holstein dairy cows producing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service