Skip to Content
Merck
All Photos(1)

Documents

28-4050

Sigma-Aldrich

Sodium salicylate

SAJ first grade, ≥99.5%

Synonym(s):

2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt, Sodium 2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COONa
CAS Number:
Molecular Weight:
160.10
Beilstein:
3732792
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

<0.1 hPa ( 25 °C)

Assay

≥99.5%

form

crystalline

availability

available only in Japan

pH

6.85 (26.3 °C)

mp

>300 °C (lit.)

SMILES string

[Na+].Oc1ccccc1C([O-])=O

InChI

1S/C7H6O3.Na/c8-6-4-2-1-3-5(6)7(9)10;/h1-4,8H,(H,9,10);/q;+1/p-1

InChI key

ABBQHOQBGMUPJH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

208.9 °F - Pensky-Martens closed cup

Flash Point(C)

98.3 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rainer Amann et al.
European journal of pharmacology, 447(1), 1-9 (2002-07-11)
Aspirin (acetylsalicylic acid) is one of the most widely used drugs worldwide. It acetylates cyclooxygenases thereby irreversibly blocking the conversion of arachidonic acid to prostanoids. Biotransformation of aspirin yields salicylate, a compound that possesses similar anti-inflammatory potency as aspirin but
Yan-Yan Su et al.
PloS one, 7(10), e46969-e46969 (2012-10-17)
Sodium salicylate (NaSal), an aspirin metabolite, can cause tinnitus in animals and human subjects. To explore neural mechanisms underlying salicylate-induced tinnitus, we examined effects of NaSal on neural activities of the medial geniculate body (MGB), an auditory thalamic nucleus that
Jos J Eggermont
Hearing research, 295, 140-149 (2012-02-15)
Animal models of tinnitus require a behavioral correlate thereof. Various conditioned response methods and gap-startle reflex methods are in use and the outcomes generally correspond with putative electrophysiological substrates of tinnitus. However, for salicylate-induced tinnitus there is discordance between the
Qian Zhang et al.
Acta biomaterialia, 8(3), 1316-1322 (2011-12-14)
Poly(amidoamine) (PAMAM) dendrons were prepared from hollow fiber membranes (HFM) consisting of bromomethylated poly(2,6-dimethyl-1,4-phenylene oxide) (BPPO) in a stepwise manner. The prepared HFM were characterized by Fourier transform infrared spectroscopy, elemental analysis, and scanning electron microscopy. The drug loading efficiency
Yutaka Sumino et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 85(4 Pt 2), 046307-046307 (2012-06-12)
The dynamics of a viscoelastic Maxwell fluid is studied in a partially filled cylinder rotating around a horizontal axis. At low rotational velocity, the fluid behaves in the same manner as a viscous fluid. A thin fluid film is pulled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service