07-0680
D-(+)-Glucose
SAJ special grade, ≥98.0%
Synonym(s):
Dextrose
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724615
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Recommended Products
grade
SAJ special grade
Assay
≥98.0%
form
solid
availability
available only in Japan
mp
150-152 °C (lit.)
SMILES string
OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1
InChI key
WQZGKKKJIJFFOK-DVKNGEFBSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A Azau et al.
Perfusion, 29(6), 496-504 (2014-03-13)
We hypothesized that the optimization of renal haemodynamics by maintaining a high level of mean arterial blood pressure (MAP) during cardiopulmonary bypass (CPB) could reduce the rate of acute kidney injury (AKI) in high-risk patients. In this randomized, controlled study
Andreia V Pinho et al.
PloS one, 10(6), e0128012-e0128012 (2015-06-06)
Sirtuin 1 (Sirt1) has been reported to be a critical positive regulator of glucose-stimulated insulin secretion in pancreatic beta-cells. The effects on islet cells and blood glucose levels when Sirt1 is deleted specifically in the pancreas are still unclear. This
Magnus Dalén et al.
Journal of cardiovascular translational research, 8(6), 381-384 (2015-07-08)
Endomyocardial delivery in the setting of active left ventricular assist device (LVAD) support has rarely been studied. The objective was to establish a protocol for endomyocardial injections during LVAD support without compromising mechanical circulation. LVAD implantation was performed in four
Mallory L Hammock et al.
Advanced materials (Deerfield Beach, Fla.), 26(35), 6138-6144 (2014-07-23)
Organic field-effect transistor (OFET) sensors can meet the need for portable and real-time diagnostics. An electronicreadout enzyme-linked immunosorbent assay using OFETs for the detection of a panel of three biomarkers in complex media to create a pre-eclampsia prognostic is demonstrated
Annelien Van Dalem et al.
Diabetologia, 58(12), 2753-2764 (2015-09-28)
We examined whether measures of glycaemic variability (GV), assessed by continuous glucose monitoring (CGM) and self-monitoring of blood glucose (SMBG), can complement or replace measures of beta cell function and insulin action in detecting the progression of preclinical disease to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service