Skip to Content
Merck
All Photos(1)

Key Documents

05-5870

Sigma-Aldrich

Copper(I) chloride

JIS special grade, ≥95.0%

Synonym(s):

Copper monochloride, Cuprous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl
CAS Number:
Molecular Weight:
99.00
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

JIS special grade

vapor pressure

1.3 mmHg ( 546 °C)

Assay

≥95.0%

form

solid

reaction suitability

reagent type: catalyst
core: copper

availability

available only in Japan

pH

5 (20 °C, 50 g/L)

bp

1490 °C (lit.)

mp

430 °C (lit.)

storage temp.

2-8°C

SMILES string

Cl[Cu]

InChI

1S/ClH.Cu/h1H;/q;+1/p-1

InChI key

OXBLHERUFWYNTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Shows unique character as an initiator of radical reactions such as the hydrostannation of α,β-unsaturated ketones.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Fan Zhao et al.
Journal of colloid and interface science, 448, 380-388 (2015-03-11)
Here we describe the development of versatile antifouling polyethersulfone (PES) filtration membranes modified via surface grafting of zwitterionic polymers from a reactive amphiphilic copolymer additive. Amphiphilic polyethersulfone-block-poly(2-hydroxyethyl methacrylate) (PES-b-PHEMA) was beforehand designed and used as the blending additive of PES
Xiaoliang Xu et al.
Organic letters, 12(5), 897-899 (2010-02-04)
Promoted by CuCl/CCl(4), a variety of sulfonyl azides and tertiary amines were successfully coupled to give sulfonyl amidine derivatives in good to excellent yields. A possible mechanism for this reaction is discussed.
Takayoshi Arai et al.
The Journal of organic chemistry, 73(13), 4903-4906 (2008-06-03)
A catalytic asymmetric Henry reaction has been developed with use of a sulfonyldiamine-CuCl complex as a catalyst. A series of new binaphthyl-containing sulfonyldiamine ligands (2a-h) were readily synthesized in two steps starting from commercially available chiral 1,2-diamines. The (R,R)-diamine-(R)-binaphthyl ligand
Yew-Foon Tan et al.
Journal of proteome research, 11(7), 3860-3879 (2012-05-12)
Plant mitochondria are highly responsive organelles that vary their metabolism in response to a wide range of chemical and environmental conditions. Quantitative proteomics studies have begun to allow the analysis of these large-scale protein changes in mitochondria. However studies of
Marie Sircoglou et al.
Journal of the American Chemical Society, 130(49), 16729-16738 (2009-06-26)
The ambiphilic triphosphine-borane ligand 1 {TPB = [o-iPr2P-(C6H4)3B} readily coordinates to all group 10 and 11 metals to afford a complete series of metal boratranes (TPB)[M] 2-8 (2: M = Ni, 3: M = Pd, 4: M = Pt, 5:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service