Skip to Content
Merck
All Photos(1)

Key Documents

05-0500

Sigma-Aldrich

Calcium carbonate

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

CP

form

powder

availability

available only in Japan

pH

8

density

2.93 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Azizur Rahman et al.
PloS one, 8(3), e58781-e58781 (2013-03-26)
An organic matrix consisting of a protein-polysaccharide complex is generally accepted as an important medium for the calcification process. While the role this "calcified organic matrix" plays in the calcification process has long been appreciated, the complex mixture of proteins
Jane A Cauley et al.
Journal of women's health (2002), 22(11), 915-929 (2013-10-18)
Clinical outcomes of the Women's Health Initiative (WHI) calcium plus vitamin D supplementation trial have been reported during 7.0 years of active intervention. We now report outcomes 4.9 years after the intervention stopped and cumulative findings. Postmenopausal women (N=36,282) were
Sophie A Jamal et al.
Lancet (London, England), 382(9900), 1268-1277 (2013-07-23)
Phosphate binders (calcium-based and calcium-free) are recommended to lower serum phosphate and prevent hyperphosphataemia in patients with chronic kidney disease, but their effects on mortality and cardiovascular outcomes are unknown. We aimed to update our meta-analysis on the effect of
Christopher T Parsons et al.
Environmental pollution (Barking, Essex : 1987), 178, 254-263 (2013-04-17)
Arsenic contamination of floodplain soils is extensive and additional fresh arsenic inputs to the pedosphere from human activities are ongoing. We investigate the cumulative effects of repetitive soil redox cycles, which occur naturally during flooding and draining, on a calcareous
Julyan H E Cartwright et al.
Angewandte Chemie (International ed. in English), 51(48), 11960-11970 (2012-11-06)
Although the polymorphism of calcium carbonate is well known, and its polymorphs--calcite, aragonite, and vaterite--have been highly studied in the context of biomineralization, polyamorphism is a much more recently discovered phenomenon, and the existence of more than one amorphous phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service