Skip to Content
Merck
All Photos(1)

Key Documents

SCC042

Sigma-Aldrich

TC28a2 Human Chondrocyte Cell Line

Human

Synonym(s):

T/C-28a2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

product name

TC28a2 Human Chondrocyte Cell Line, TC28a2 Human Chondrocyte Cell Line is widely used as a model cell line for studying normal and pathological cartilage repair mechanisms related to chondrocyte biology and physiology.

biological source

human

Quality Level

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

General description

The chondrocyte is the only specialized cell type present in articular cartilage. They produce secretions to support and repair the cartilage matrix. Human chondrocytes in mature articular cartilage are post-mitotic and terminally differentiated cells. Studies on human chondrocytes have been hampered by the difficulty in obtaining sufficient numbers of primary human chondrocytes from a single joint and by variabilities among donors due to factors such as age and medical conditions.

T/C-28a2 cell line was established by transfecting primary cultures (day 5) of costal cartilage from a 15-year-old female with a retroviral vector expressing simian virus SV40 large T antigen.

References:
1. Goldring, M.B, Birkhead, J.R., Suen, L.F., Yamin, R., Mizuno, S., Glowacki, J., Arbiser, J.L, Apperley, J.F. (1994) Interleukin-1 beta-modulated gene expression in immortalized human chondrocytes. J. Clin. Invest 94(6): 2307-2316.
2. Claassen, H., Schicht, M., Brandt, J., Reuse, K., Schadlich, R., Goldring, M.B., Guddat, S.S., Thate, A., Paulsen, F. C-28/I2 and T/C-28a2 chondrocytes as well as human primary articular chondrocytes express hormone and insulin receptors- Useful cells in study of cartilage metabolism Ann Anat. 193(1): 23-29.
3. Goldring, M.B. (2004) Culture of immortalized chondrocytes and their use as models of chondrocyte function. Methods Mol. Med. 100: 37-52.

Cell Line Description

Chondrocyte Cells

Application

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Quality

• Each vial contains ≥ 1X106 viable cells.
• Cells are tested by PCR and are negative for HPV-16, HPV-18, Hepatitis A, B, C and HIV-1 & 2 viruses.
• Cells are negative for mycoplasma contamination.
• Each lot of cells are genotyped by STR analysis to verify the unique identity of the cell line.

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piotr Kuropka et al.
Materials (Basel, Switzerland), 14(24) (2021-12-25)
Graphene has been one of the most tested materials since its discovery in 2004. It is known for its special properties, such as electrical conductivity, elasticity and flexibility, antimicrobial effect, and high biocompatibility with many mammal cells. In medicine, the
Rocío Mato-Basalo et al.
International journal of molecular sciences, 22(7) (2021-04-04)
Mesenchymal stem cells have an important potential in the treatment of age-related diseases. In the last years, small extracellular vesicles derived from these stem cells have been proposed as cell-free therapies. Cellular senescence and proinflammatory activation are involved in the
Junquera López-Seijas et al.
Molecular therapy. Nucleic acids, 32, 302-317 (2023-04-25)
Senescence is a process characterized by a prolonged irreversible cell-cycle arrest. The accumulation of senescent cells in tissues is related to aging and to the development of age-related diseases. Recently, gene therapy has emerged as a powerful tool for treating
J Víctor Álvarez et al.
Pharmaceutics, 11(10) (2019-10-17)
Mucopolysaccharidosis IVA (Morquio A) is a rare inherited metabolic disease caused by deficiency of the lysosomal enzyme N-acetylgalatosamine-6-sulfate-sulfatase (GALNS). Until now, treatments employed included hematopoietic stem cell transplantation and enzyme replacement therapy (ERT); the latter being the most commonly used
Mary B Goldring
Methods in molecular medicine, 100, 37-52 (2004-07-29)
Immortalization of chondrocytes increases life span and proliferative capacity but does not necessarily stabilize the differentiated phenotype. Expansion of chondrocyte cell lines in continuous monolayer culture may result in the loss of phenotype, particularly if high cell density is not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service