Skip to Content
Merck
All Photos(2)

Key Documents

PEZGS0816

Millipore

Millicell® EZ Slide

8-well glass chamber slide, 0.7 cm2/well, 16/pk, sterile

Synonym(s):

8 well glass slide, Chamber slides

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NB.22
Pricing and availability is not currently available.

material

polypropylene wells
soda-lime glass slide

Quality Level

sterility

ethylene oxide treated
sterile

feature

Raised corner ridges on the lid facilitate stacking of multiple units

packaging

box of 16 units (4 trays, 4 units/tray)

manufacturer/tradename

Millicell®

technique(s)

cell based assay: suitable
cell culture | mammalian: suitable
immunocytochemistry: suitable

H

13.9 mm , excluding lid
16.2 mm , including lid

L × W × H

75 mm × 25 mm × 1 mm, slide dimensions

L

77.8 mm

W

33.8 mm

size

8 wells

well surface area

0.7 cm2

well volume

200-700 μL

shipped in

ambient

General description

Millicell® EZ Slide allows the user to grow, fix, and stain cells directly on a microscope slide. There is no need to disassemble the unit or transfer cells from one device to another prior to analysis. The polypropylene wells are resistant to most fixatives used, so the cells can be grown on the slide and stained right in the well. Once stained, the microscope slide can be easily removed by pressing down the four side tabs of the unit and lifting the wells off of the slide. This easy slide removal reduces the risk of damaging the cells and no special tools are required to remove the wells from the slide. Raised corner ridges on the cover facilitate stacking of multiple units in the incubator, or they can be incubated in the tray supplied with the units.

Preparation Note

  1. Culture, fix, and stain the cells according to preferred protocol.
  2. To access the microscope slide, hold the Millicell® EZ SLIDE down on a flat surface and break the four tabs by pushing them down with the thumb. Remove the cover. Hold the slide down and lift the wells upward to remove. Remove slide from holder.

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lulu Hou et al.
Investigative ophthalmology & visual science, 61(5), 7-7 (2020-05-12)
To investigate the effect and mechanism of Agrin on limbal stem cell proliferation and corneal wound healing. Limbal stem cells were isolated and treated with different concentrations of Agrin. CCK-8 and cell proliferation markers (Ki67 and pH3) were detected to
Anna A Kondratova et al.
The EMBO journal, 39(11), e101573-e101573 (2020-04-24)
High expression of 2',5'-oligoadenylate synthetase 1 (OAS1), which adds AMP residues in 2',5' linkage to a variety of substrates, is observed in many cancers as a part of the interferon-related DNA damage resistance signature (IRDS). Poly(ADP-ribose) (PAR) is rapidly synthesized
Man Ying Wong et al.
Journal of neuroinflammation, 17(1), 192-192 (2020-06-20)
Genome-wide association studies of Alzheimer's disease (AD) have implicated pathways related to lipid homeostasis and innate immunity in AD pathophysiology. However, the exact cellular and chemical mediators of neuroinflammation in AD remain poorly understood. The oxysterol 25-hydroxycholesterol (25-HC) is an
Guang-Chun Wang et al.
Journal of inflammation (London, England), 17, 2-2 (2020-01-09)
The relationship between chronic prostatitis/chronic pelvic pain syndrome (CP/CPPS) and erectile dysfunction (ED) has been shown in many studies. However, the specific mechanism remains unclear. This study was to investigate the corpus cavernosum smooth muscle cell function and phenotype transformation
Sarah Rank Rønnow et al.
Respiratory research, 21(1), 108-108 (2020-05-10)
Idiopathic pulmonary fibrosis (IPF) is a rapidly progressing disease with challenging management. To find novel effective therapies, better preclinical models are needed for the screening of anti-fibrotic compounds. Activated fibroblasts drive fibrogenesis and are the main cells responsible for the

Articles

High titer lentiviral particles for LC3 variants used for live cell analysis of cellular autophagy.

High titer lentiviral particles including beta-actin, alpha-tubulin and vimentin used for live cell analysis of cytoskeleton structure proteins.

High titer lentiviral particles including beta-actin, alpha-tubulin and vimentin used for live cell analysis of cytoskeleton structure proteins.

High titer lentiviral particles including beta-actin, alpha-tubulin and vimentin used for live cell analysis of cytoskeleton structure proteins.

See All

Questions

1–4 of 4 Questions  
  1. What size of cover slip is compatible with Millicell EZ Slides?

    1 answer
    1. The compatible coverslips for Millicell EZ Slides are standard rectangular glass measuring 24 x 50 mm. An example would be C8181.

      Helpful?

  2. What are the dimensions of this product? What is the corresponding cover glass product that you would recommend I purchase with this?

    1 answer
    1. The following specifications are associated with this product:
      Property | 4 well | 8 well
      Recommended media volume | 0.5 - 1.7 mL | 0.2 - 0.7 mL
      Length | 77.8 mm | 77.8 mm
      Width | 33.8 mm | 33.8 mm
      Height (excluding cover) | 13.9 mm | 13.9 mm
      Height (including cover) | 16.2 mm | 16.2 mm
      Growing area per well | 1.7 cm^2 | 0.7 cm^2
      Slide dimensions | 25 x 75 x 1mm | 25 x 75 x 1 mm

      The recommendation for a coverslips is the following below:
      24 x 50 glass coverslips
      https://www.sigmaaldrich.com/product/sigma/cls2975245

      Helpful?

  3. What are the dimensions of each well?

    1 answer
    1. See the image below for complete dimensions of this product: (attach image)

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service