Skip to Content
Merck
All Photos(1)

Key Documents

MYSK10025

Millipore

Yeast and Mold Test Kit

for yeasts, for molds

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.74

application(s)

food and beverages
personnel monitoring
surface monitoring
water monitoring

Quality Level

suitability

molds
yeasts

General description

The Swab Test Kit is a self-contained system, incorporating Yeast and mold Samplers and units containing 18mL of sterile, phosphate buffer into which are fitted swab assemblies. The system is designed for use as swab tests for measuring yeast and mold levels on flat or irregular surfaces. The Yeast and Mould Swab Test Kit enables a fast and easy Yeast and Mould monitoring in food and beverages, dental unit water, dialysate, pharmaceuticals and cosmetics. The buffer units contain neutralizing agents to counteract the adverse effect of any residual chlorine or quaternary ammonium compounds that may be present on surfaces after sanitization. The kit contains 25 x dip tester + 25 x swabs

Application

The Yeast and Mould Swab Test Kit enables a fast and easy Yeast and Mould monitoring in food and beverages, dental unit water, dialysate, clinical lab, pharmaceuticals and cosmetics.

Analysis Note

Yeast colonies appear satiny, opaque, white coloured or may turn green over time. Mould colonies appear white, green or brown/black and filamentous. Bacterial colonies may appear but are usually smaller and more glistening and transparent than the yeast colonies.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service