Skip to Content
Merck
All Photos(1)

Documents

MAB3318

Sigma-Aldrich

Anti-TIMP-3 Antibody, a.a. 170-188, clone 136-13H4

clone 136-13H4, Chemicon®, from mouse

Synonym(s):

Tissue Inhibitor of Metalloproteinase-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

136-13H4, monoclonal

species reactivity

rabbit, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TIMP3(7078)

Specificity

Specifically reacts with human TIMP-3. This is a purified mouse monoclonal antibody to an oligopeptide of SWYRGWAPPDKSIINATDP (residue 170-188) on human tissue inhibitor of metalloproteinases-3 (human TIMP-3).
MAB3318 cross-reacts with rabbit TIMP-3, but does not react with human TIMP-1 and human TIMP-2.

Immunogen

Epitope: a.a. 170-188
Oligopeptide of SWYRGWAPPDKSIINATDP (residue 170-188) on human tissue inhibitor of metalloproteinases-3.

Application

Anti-TIMP-3 Antibody, a.a. 170-188, clone 136-13H4 is an antibody against TIMP-3 for use in IH(P) & WB.
Immunoblotting: 2 ug/mL

Immunohistochemistry (Paraffin):10ug/mL

Immunohistochemistry (Frozen): 2 ug/mL
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs

Target description

26 kDa

Physical form

Format: Purified
Liquid in 0.1 M sodium phosphate buffer, pH 7.0 containing 2% protease free bovine Serum albumin.
Protein A purified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Breast carcinoma tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Manufactured by Daiichi Fine Chemical Co., Ltd

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Wen Su et al.
Cell death & disease, 10(11), 793-793 (2019-10-19)
The tissue inhibitor of metalloproteinase-3 (TIMP3) is the only member of the TIMP family that binds to the extracellular matrix and suppresses cancer cell growth, angiogenesis, migration, and invasion. However, whether the abnormal expression and promoter methylation of TIMP3 facilitates
S Jane Darnton et al.
International journal of cancer, 115(3), 351-358 (2005-02-03)
Amongst involvement in diverse physiological and pathological processes, TIMP-3 may have an important role in tumour development, growth and metastasis by interaction with metalloproteases in the extracellular matrix. We studied the role and prognostic effect of TIMP-3 in esophageal adenocarcinoma
Lluis Catasus et al.
Histopathology, 62(4), 632-641 (2013-02-06)
Expression of tissue inhibitor of metalloproteinases-3 (TIMP-3) has been found to be decreased in several types of cancer by promoter gene hypermethylation. However, little is known regarding the silencing effect of TIMP3 promoter hypermethylation on gene and protein expression in
Qinglei Li et al.
Reproduction (Cambridge, England), 128(5), 555-564 (2004-10-29)
The matrix metalloproteinases (MMPs) and their tissue inhibitors (TIMPs) are potential regulators of the focalized extracellular matrix degradation required for ovulation. The objectives of the present study were to determine localization and temporal regulation of TIMP-3 and TIMP-4 mRNA and
Classical macrophage activation up-regulates several matrix metalloproteinases through mitogen activated protein kinases and nuclear factor-?B.
Huang, WC; Sala-Newby, GB; Susana, A; Johnson, JL; Newby, AC
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service