Skip to Content
Merck
All Photos(1)

Key Documents

CBL1304

Sigma-Aldrich

Anti-Integrin α4 Antibody, clone PS/2

clone PS/2, Chemicon®, from rat

Synonym(s):

CD49d, VLA-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PS/2, monoclonal

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA4(3676)
mouse ... Itga4(16401)

General description

Virtually all lymphoid and myeloid cells in bone marrow are reactive with the anti-VLA-4 antibody. VLA-4 is absent or minimally detectable on stromal cells. Myeloid cells maintained in Dexter-type long-term cultures of murine bone marrow are positive, as are lymphoid cells from Whitlock-White cultures.

Specificity

Clone PS/2 recognises the murine alpha4 chain of the VLA-4 integrin heterodimer, 150 kDa. The antibody also reacts with human VLA-4.
Reactivity with other species has not been confirmed.

Application

Anti-Integrin α4 Antibody, clone PS/2 detects level of Integrin α4 & has been published & validated for use in FC, IF, IP, IH.
Flow cytometry: 2 μg per 106 cells

Immunohistochemistry on frozen tissue sections

Immunoprecipitation

Cell adhesion assays

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Linkage

Replaces: 04-1131

Physical form

Format: Purified
Purified rat IgG2bκ in 0.01 M phosphate buffer, pH 7.1 containing no preservatives.

Storage and Stability

Maintain at -20ºC for up to 12 months from date of receipt.Upon first use, aliquot into smaller quantities to avoid additional freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ursula Norman et al.
European journal of immunology, 38(5), 1368-1380 (2008-04-17)
CD4(+) T helper (Th1 and Th2) cell localization to a site of inflammation is important for the development, maintenance and regulation of an immune response. The factors that regulate Th1 and Th2 cell recruitment into tissue are not fully understood.
Julia C Gutjahr et al.
Haematologica, 106(8), 2102-2113 (2020-07-04)
Adhesive properties of leukemia cells shape the degree of organ infiltration and the extent of leukocytosis. CD44 and the integrin VLA-4, a CD49d/CD29 heterodimer, are important factors of progenitor cell adhesion in bone marrow (BM). Here, we report their cooperation
Xuan Jiang et al.
Nature communications, 8(1), 128-128 (2017-07-27)
Hematopoietic stem and progenitor cells arise from the vascular endothelium of the dorsal aorta and subsequently switch niche to the fetal liver through unknown mechanisms. Here we report that vascular endothelium-specific deletion of mouse Drosha (Drosha cKO), an enzyme essential
Integrins and cAMP mediate netrin-induced growth cone collapse.
Lemons, ML; Abanto, ML; Dambrouskas, N; Clements, CC; Deloughery, Z; Garozzo, J; Condic, ML
Brain Research null
Michael C Schmid et al.
Nature communications, 13(1), 1768-1768 (2022-04-03)
Myeloid cells play key roles in cancer immune suppression and tumor progression. In response to tumor derived factors, circulating monocytes and granulocytes extravasate into the tumor parenchyma where they stimulate angiogenesis, immune suppression and tumor progression. Chemokines, cytokines and interleukins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service