Skip to Content
Merck
All Photos(1)

Key Documents

AB1532P

Sigma-Aldrich

Anti-Neurotrophin 3 Antibody

Chemicon®, from rabbit

Synonym(s):

NT3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, chicken

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NTF3(4908)

Specificity

Neurotrophin 3 (NT3). Less than 0.1% cross reactivity with recombinant human BDNF, recombinant human NT4 and mouse NGF.

Immunogen

An 11 amino acid peptide corresponding to the amino-terminal of mouse Neurotrophin 3 (NT3) coupled to BSA. The sequence is H2N-YAEHKSHRGEY-.

Application

Detect Neurotrophin 3 using this Anti-Neurotrophin 3 Antibody validated for use in WB, IH.
Immunohistochemistry: 1-10 μg/mL (see suggested protocol).


Immunoblotting:1-10 μg/mL


Inhibition of biological activity in vitro: 1-10 μg/mL


In vivo animal studies


Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Physical form

Affinity purified immunoglobulin. Lyophilized from PBS, no preservatives. Reconstitute to desired volume with sterile distilled water. Suggested concentration 0.5-1 mg/mL.

Storage and Stability

Maintain at -20 to -70°C in undiluted aliquots for up to one year. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huan Liu et al.
Experimental neurology, 241, 125-137 (2012-12-25)
The olfactory system continuously incorporates new neurons into functional circuits throughout life. Axons from olfactory sensory neurons (OSNs) in the nasal cavity synapse on mitral, tufted and periglomerular (PG) cells in the main olfactory bulb, and low levels of turnover

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service