Skip to Content
Merck
All Photos(1)

Key Documents

8.07340

Sigma-Aldrich

Hexachloroplatinic(IV) acid hexahydrate

~40% Pt, for synthesis

Synonym(s):

Hexachloroplatinic(IV) acid hexahydrate (~40% Pt), Chloroplatinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2(PtCl6) · 6H2O
CAS Number:
Molecular Weight:
517.90
MDL number:
UNSPSC Code:
12352301
EC Index Number:
241-010-7
NACRES:
NA.22

product name

Hexachloroplatinic(IV) acid hexahydrate (~40% Pt), for synthesis

Quality Level

100
200

form

solid

concentration

39-41% Pt

mp

150 °C

solubility

water: soluble

density

4 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/6Cl.6H2O.Pt/h;;;;;;6*1H2;/q;;;;;;;;;;;;-2/p+2

InChI key

YYOLYBUNSBKNLR-UHFFFAOYSA-P

Application

Hexachloroplatinic(IV) acid hexahydrate can be used as a platinum precursor to synthesize:
  • Pt/C catalyst for polymer electrolyte membrane fuel cell applications.
  • Pt/TiO2 catalyst to study the photocatalytic degradation pathway of methylene blue.

Analysis Note

Assay (of Pt): 39.0 - 41.0 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Platinum catalyst supported on mesoporous carbon for PEMFC
Ambrosio EP, et al.
International Journal of Hydrogen Energy, 33(12), 3142-3145 (2008)
The effect of Pt on the photocatalytic degradation pathway of methylene blue over TiO2 under ambient conditions
Yu Zhiqiang and Chuang S SC
Applied Catalysis. B, Environmental, 83(3-4), 277-285 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service