Skip to Content
Merck
All Photos(1)

Key Documents

565750

Sigma-Aldrich

γ-Secretase Inhibitor I

The γ-Secretase Inhibitor I controls the biological activity of γ-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

γ-Secretase Inhibitor I, Z-LLNle-CHO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H41N3O5
Molecular Weight:
475.62
UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

DMSO: 10 mg/mL
50% acetic acid: soluble

shipped in

ambient

storage temp.

−20°C

General description

Cell permeable. Inhibits γ-secretase, a protease that cleaves the Amyloid Precursor Protein (APP) to generate β-amyloid protein. Inhibition of this enzyme is believed to aid in the prevention of Alzheimer’s disease.
Inhibits γ-secretase, a protease that cleaves the amyloid precursor protein (APP). Inhibition of this enzyme is believed to aid in the prevention of Alzheimer′s disease.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
γ-secretase
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Leu-Leu-Nle-CHO (Nle = Norleucine)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Skovronsky, D.M., et al. 2000. Biochemistry 39, 810.
Hartmann, T., et al. 1997. Nat. Med. 3, 1016.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Shreoshi Sengupta et al.
eLife, 11 (2022-06-02)
Cancer stem cells (CSCs) alone can initiate and maintain tumors, but the function of non-cancer stem cells (non-CSCs) that form the tumor bulk remains poorly understood. Proteomic analysis showed a higher abundance of the extracellular matrix small leucine-rich proteoglycan fibromodulin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service