Skip to Content
Merck
All Photos(1)

Key Documents

48-621MAG

Millipore

MILLIPLEX® DNA Damage/Genotoxicity Magnetic Bead Panel - Cell Signaling Multiplex Assay

Synonym(s):

Cell Signaling DNA Damage Kit, DNA Damage Bead Panel, Genotoxicity Magnetic Beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

species reactivity

mouse, human, rat

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

The MILLIPLEX® MAP 7-plex DNA Damage/Genotoxicity Magnetic Bead Kit (Cat. No. 48-621MAG) is a premixed kit to be used for the simultaneous quantification of the following 7 analytes in cell lysates:• ATR (total)• Chk1 (Ser345)• Chk2 (Thr68)• H2A.X (Ser139)• MDM2 (total)• p21 (total)• p53 (Ser15)

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

Intracellular Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous relative quantitation of multiple phosphorylation and total pathway proteins in tissue and cell lysate samples. Compare Multiplexing results to those of Western blotting.An overnight (4°C) incubation is recommended for best results.This assay requires 25 μL diluted cell lysate per well.This kit must be run using Assay Buffer 1 (provided).The suggested working range of protein concentration for the assay is 1 to 25 μg of total protein/well (25 μL/well at 40 to 1,000 μg/mL).Analytes available:ATR (Total);Chk1 (Ser345);Chk2 (Thr68);H2A.X (Ser139);MDM2 (total);p21 (Total);p53 (Ser15)

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

MILLIPLEX® genotoxicity assays enable high-throughput measurement of DNA damage, reducing time and cost.

MILLIPLEX® genotoxicity assays enable high-throughput measurement of DNA damage, reducing time and cost.

MILLIPLEX® genotoxicity assays enable high-throughput measurement of DNA damage, reducing time and cost.

MILLIPLEX® genotoxicity assays enable high-throughput measurement of DNA damage, reducing time and cost.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service